Learn More
Meso Scale Discovery UPLEX MSE CD40/TNFRSF5 ASY384
SDP
Catalog No. 5000081212
Encompass
The U-PLEX Mouse CD40/TNFRSF5 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD40 and TNFRSF5 in mouse samples It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 10 L of sample per well and the kit includes a plate detection antibodies read buffer wash buffer diluent and calibrators It can detect CD40 and TNFRSF5 simultaneously in serum plasma tissue lysates and cell culture supernatants
Catalog No. 50-000-81212 Supplier Meso Scale Discovery Supplier No. K252D0K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse CD40/TNFRSF5 384-well Assay Kit from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD40 and TNFRSF5 in mouse samples It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 10 L of sample per well and the kit includes a plate detection antibodies read buffer wash buffer diluent and calibrators It can detect CD40 and TNFRSF5 simultaneously in serum plasma tissue lysates and cell culture supernatants