Learn More
Meso Scale Discovery UPLEX MSE CD40/TNFRSF5 ASY384
SDP
Catalog No. 5000081254
Encompass
The U-PLEX Mouse CD40/TNFRSF5 384-well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring CD40 and TNFRSF5 in mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation from serum plasma and cell culture supernatants The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG and calibrators for standard curves The signal offers a wide dynamic range and is compatible with automated systems for high throughput
Catalog No. 50-000-81254 Supplier Meso Scale Discovery Supplier No. K252D0K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse CD40/TNFRSF5 384-well Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring CD40 and TNFRSF5 in mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantitation from serum plasma and cell culture supernatants The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG and calibrators for standard curves The signal offers a wide dynamic range and is compatible with automated systems for high throughput