Learn More
Meso Scale Discovery UPLEX MSE CPeptide ASY
SDP
Catalog No. 5000081497
Encompass
The Meso Scale Discovery Mouse C-Peptide Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures C-peptide levels in mouse serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume This assay allows for the accurate quantification of C-peptide an important biomarker for diabetes research and drug discovery With a rapid 2 5 hour protocol minimal sample dilution requirements and compatibility with common sample matrices this kit enables robust and reproducible analysis of C-peptide levels
Catalog No. 50-000-81497 Supplier Meso Scale Discovery Supplier No. K1526JK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse C-Peptide Singleplex Assay Kit is a singleplex immunoassay that quantitatively measures C-peptide levels in mouse serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume This assay allows for the accurate quantification of C-peptide an important biomarker for diabetes research and drug discovery With a rapid 2 5 hour protocol minimal sample dilution requirements and compatibility with common sample matrices this kit enables robust and reproducible analysis of C-peptide levels