Learn More
Meso Scale Discovery UPLEX MSE CPeptide ASY384well
SDP
Catalog No. 5000078971
Encompass
The Meso Scale Discovery U-PLEX Mouse C-Peptide 384-Well Assay Kit (K2526JK) is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a sensitivity of 0 5 pg/mL and a working range of 0 5-1000 pg/mL Results are obtained in 3 5 hours using an MSD instrument capable of reading 384-well plates This assay allows high-throughput quantification of C-peptide levels in mouse samples
Catalog No. 50-000-78971 Supplier Meso Scale Discovery Supplier No. K2526JK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse C-Peptide 384-Well Assay Kit (K2526JK) is an electrochemiluminescent immunoassay for the quantitative measurement of C-peptide in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent The assay has a sensitivity of 0 5 pg/mL and a working range of 0 5-1000 pg/mL Results are obtained in 3 5 hours using an MSD instrument capable of reading 384-well plates This assay allows high-throughput quantification of C-peptide levels in mouse samples