Learn More
Meso Scale Discovery UPLEX MSE CPeptide ASY384well
SDP
Catalog No. 5000081273
Encompass
The Meso Scale Discovery U-PLEX Mouse C-Peptide 384-Well Assay Kit (K2526JK) is an electrochemiluminescent immunoassay for quantifying C-peptide in mouse serum plasma and cell culture supernatants It includes a 384-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators and diluent Sensitivity is 0 03 ng/mL with a range of 0 03-1000 ng/mL The sandwich immunoassay format takes 3 5 hours The kit provides reagents for 3 x 384-well plates enabling high-throughput sensitive and accurate C-peptide measurement in mouse samples
Catalog No. 50-000-81273 Supplier Meso Scale Discovery Supplier No. K2526JK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse C-Peptide 384-Well Assay Kit (K2526JK) is an electrochemiluminescent immunoassay for quantifying C-peptide in mouse serum plasma and cell culture supernatants It includes a 384-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators and diluent Sensitivity is 0 03 ng/mL with a range of 0 03-1000 ng/mL The sandwich immunoassay format takes 3 5 hours The kit provides reagents for 3 x 384-well plates enabling high-throughput sensitive and accurate C-peptide measurement in mouse samples