Learn More
Meso Scale Discovery UPLEX MSE CPeptide ASYQP
SDP
Catalog No. 5000079258
Encompass
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a multiplex immunoassay kit for quantifying mouse C-peptide in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision with minimal interference The assay requires 50 L or less sample volume and is compatible with MSD instruments like SECTOR Imager and QuickPlex SQ 120 The kit includes necessary components for generating a standard curve and validating performance enabling high-throughput reproducible analysis of mouse C-peptide levels
Catalog No. 50-000-79258 Supplier Meso Scale Discovery Supplier No. K1526JK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse C-Peptide Assay is a multiplex immunoassay kit for quantifying mouse C-peptide in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision with minimal interference The assay requires 50 L or less sample volume and is compatible with MSD instruments like SECTOR Imager and QuickPlex SQ 120 The kit includes necessary components for generating a standard curve and validating performance enabling high-throughput reproducible analysis of mouse C-peptide levels