Learn More
Meso Scale Discovery UPLEX MSE Eotaxin ASY
SDP
Catalog No. 5000078710
Encompass
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that allows simultaneous quantification of mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection is 0 06 pg/mL This assay kit includes a 96-well MULTI-ARRAY Mouse Eotaxin Calibrator and all the reagents needed to perform the assay
Catalog No. 50-000-78710 Supplier Meso Scale Discovery Supplier No. K152UDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that allows simultaneous quantification of mouse Eotaxin in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection is 0 06 pg/mL This assay kit includes a 96-well MULTI-ARRAY Mouse Eotaxin Calibrator and all the reagents needed to perform the assay