Learn More
Meso Scale Discovery UPLEX MSE Eotaxin ASYQP
SDP
Catalog No. 5000078709
Encompass
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that quantitatively measures Eotaxin in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies and calibrators The assay has high sensitivity and broad dynamic range requires low sample volume and provides results in as little as 3 5 hours With minimal hands-on time required this assay allows for rapid reproducible quantification of mouse Eotaxin from various sample types
Catalog No. 50-000-78709 Supplier Meso Scale Discovery Supplier No. K152UDK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Eotaxin Assay Kit is a multiplex immunoassay that quantitatively measures Eotaxin in mouse biological samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient all-in-one kit containing plates coated with capture antibodies detection antibodies and calibrators The assay has high sensitivity and broad dynamic range requires low sample volume and provides results in as little as 3 5 hours With minimal hands-on time required this assay allows for rapid reproducible quantification of mouse Eotaxin from various sample types