Learn More
Meso Scale Discovery UPLEX MSE Ghrelin act ASY384
SDP
Catalog No. 5000078972
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex kit includes a plate coated with capture antibodies for active ghrelin along with detection antibodies and calibrators It allows rapid and accurate quantification of active ghrelin levels with as little as 25 L of sample
Catalog No. 50-000-78972 Supplier Meso Scale Discovery Supplier No. K2526KK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex kit includes a plate coated with capture antibodies for active ghrelin along with detection antibodies and calibrators It allows rapid and accurate quantification of active ghrelin levels with as little as 25 L of sample