Learn More
Meso Scale Discovery UPLEX MSE Ghrelin act ASYQP
SDP
Catalog No. 5000079254
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit measures active ghrelin in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Sensitivity is 1 3 pg/mL with a quantification limit of 2500 pg/mL for serum/plasma This kit allows high-throughput quantification of active ghrelin in mouse samples
Catalog No. 50-000-79254 Supplier Meso Scale Discovery Supplier No. K1526KK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit measures active ghrelin in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Sensitivity is 1 3 pg/mL with a quantification limit of 2500 pg/mL for serum/plasma This kit allows high-throughput quantification of active ghrelin in mouse samples