Learn More
Meso Scale Discovery UPLEX MSE Ghrelin active ASY
SDP
Catalog No. 5000080021
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Active Assay Kit measures active ghrelin in mouse samples like serum plasma and tissue homogenates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and necessary reagents Results are read on MSD s SECTOR Imager systems offering a rapid high-throughput method to quantify active ghrelin levels
Catalog No. 50-000-80021 Supplier Meso Scale Discovery Supplier No. K1526KK1
The Meso Scale Discovery U-PLEX Mouse Ghrelin Active Assay Kit measures active ghrelin in mouse samples like serum plasma and tissue homogenates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and necessary reagents Results are read on MSD s SECTOR Imager systems offering a rapid high-throughput method to quantify active ghrelin levels