Learn More
Meso Scale Discovery UPLEX MSE Ghrelin active ASYQP
SDP
Catalog No. 5000078481
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of active ghrelin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for serum/plasma samples
Catalog No. 50-000-78481 Supplier Meso Scale Discovery Supplier No. K1526KK21
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of active ghrelin The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for serum/plasma samples