Learn More
Meso Scale Discovery UPLEX MSE Ghrelin active ASYQP
SDP
Catalog No. 5000081328
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin levels in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for detection on the MESO QuickPlex SQ 120 instrument This assay allows rapid accurate quantification of active ghrelin levels from small sample volumes
Catalog No. 50-000-81328 Supplier Meso Scale Discovery Supplier No. K1526KK22
The Meso Scale Discovery U-PLEX Mouse Ghrelin (Active) Assay Kit is a multiplex immunoassay that measures active ghrelin levels in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer for detection on the MESO QuickPlex SQ 120 instrument This assay allows rapid accurate quantification of active ghrelin levels from small sample volumes