Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASY
SDP
Catalog No. 5000082071
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit measures total ghrelin in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 2500 pg/mL enabling accurate ghrelin quantification for mouse research
Catalog No. 50-000-82071 Supplier Meso Scale Discovery Supplier No. K1525XK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit measures total ghrelin in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 2500 pg/mL enabling accurate ghrelin quantification for mouse research