Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASY
SDP
Catalog No. 5000081325
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in mouse serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve Results are measured with an MSD instrument like the MESO QuickPlex SQ 120 aiding in the study of appetite regulation and energy metabolism in mouse models
Catalog No. 50-000-81325 Supplier Meso Scale Discovery Supplier No. K1525XK2
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in mouse serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and calibrators for a standard curve Results are measured with an MSD instrument like the MESO QuickPlex SQ 120 aiding in the study of appetite regulation and energy metabolism in mouse models