Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASY384
SDP
Catalog No. 5000079982
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total 384-Well Assay Kit (K2525XK) is a multiplex immunoassay kit for the quantitative measurement of total ghrelin in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit allows for the accurate quantification of total ghrelin levels from small sample volumes
Catalog No. 50-000-79982 Supplier Meso Scale Discovery Supplier No. K2525XK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total 384-Well Assay Kit (K2525XK) is a multiplex immunoassay kit for the quantitative measurement of total ghrelin in mouse serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibodies detection antibodies calibrators and buffers needed to run the assay The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay kit allows for the accurate quantification of total ghrelin levels from small sample volumes