Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASY384
SDP
Catalog No. 5000081215
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total 384-Well Assay Kit (K2525XK) is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of total ghrelin The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and an assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse total ghrelin with a sample volume of 25 L
Catalog No. 50-000-81215 Supplier Meso Scale Discovery Supplier No. K2525XK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total 384-Well Assay Kit (K2525XK) is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of total ghrelin in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of total ghrelin The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and an assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse total ghrelin with a sample volume of 25 L