Learn More
Meso Scale Discovery UPLEX MSE Ghrelin TTL ASYQP
SDP
Catalog No. 5000081406
Encompass
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit measures total ghrelin in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and buffers Results are read on MSD s QuickPlex SQ 120 and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method to quantify ghrelin levels
Catalog No. 50-000-81406 Supplier Meso Scale Discovery Supplier No. K1525XK22
The Meso Scale Discovery U-PLEX Mouse Ghrelin Total Assay Kit measures total ghrelin in mouse samples like serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and buffers Results are read on MSD s QuickPlex SQ 120 and analyzed with MSD Discovery Workbench software offering a rapid high-throughput method to quantify ghrelin levels