Learn More
Meso Scale Discovery UPLEX MSE GLP1 active ASY
SDP
Supplier:  Meso Scale Discovery K1526LK4
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a ruthenylated detection antibody to provide high sensitivity and a broad dynamic range The kit includes a plate detection antibody calibrator and assay diluent valid for 50 tests with a 3 5 hour protocol The assay has high specificity for active GLP-1 with no cross-reactivity to inactive GLP-1 or glucagon
Catalog No. 50-000-82380
Only null left
Add to Cart
Request a Quote