Learn More
Meso Scale Discovery UPLEX MSE GLP1 active ASYQP
SDP
Catalog No. 5000080018
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit K1526LK is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants This assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody to measure active GLP-1 levels down to 0 61 pg/mL The kit includes a 96-well plate detection antibody read buffer calibrator diluent wash buffer and calibrators to generate a standard curve This assay provides high sensitivity low sample volume requirements and a rapid 3 5 hour protocol
Catalog No. 50-000-80018 Supplier Meso Scale Discovery Supplier No. K1526LK21
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit K1526LK is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants This assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody to measure active GLP-1 levels down to 0 61 pg/mL The kit includes a 96-well plate detection antibody read buffer calibrator diluent wash buffer and calibrators to generate a standard curve This assay provides high sensitivity low sample volume requirements and a rapid 3 5 hour protocol