Learn More
Meso Scale Discovery UPLEX MSE GLP1 active ASYQP
SDP
Catalog No. 5000082201
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants This assay uses MSD GOLD technology to provide superior sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent The active GLP-1 assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL This assay allows for the specific measurement of biologically active GLP-1
Catalog No. 50-000-82201 Supplier Meso Scale Discovery Supplier No. K1526LK22
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative determination of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants This assay uses MSD GOLD technology to provide superior sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent The active GLP-1 assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL This assay allows for the specific measurement of biologically active GLP-1