Learn More
Meso Scale Discovery UPLEX MSE GLP1 active ASYQP
SDP
Catalog No. 5000082202
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a ruthenylated detection antibody to provide high sensitivity and a broad dynamic range The kit includes a plate detection antibody calibrator and assay diluent valid for 50 tests with a 3 5 hour protocol This assay is useful for studying GLP-1 biology and testing therapeutic modulation of active GLP-1 levels in preclinical mouse models
Catalog No. 50-000-82202 Supplier Meso Scale Discovery Supplier No. K1526LK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with a GLP-1 specific capture antibody and a ruthenylated detection antibody to provide high sensitivity and a broad dynamic range The kit includes a plate detection antibody calibrator and assay diluent valid for 50 tests with a 3 5 hour protocol This assay is useful for studying GLP-1 biology and testing therapeutic modulation of active GLP-1 levels in preclinical mouse models