Learn More
Meso Scale Discovery UPLEX MSE GLP1 inactive ASY
SDP
Catalog No. 5000081490
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for measuring inactive GLP-1 in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The assay detects inactive GLP-1 from 50-10 000 pg/mL and includes a plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an MSD read buffer for signal generation enabling accurate quantification of inactive GLP-1 in mouse samples
Catalog No. 50-000-81490 Supplier Meso Scale Discovery Supplier No. K1525VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for measuring inactive GLP-1 in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The assay detects inactive GLP-1 from 50-10 000 pg/mL and includes a plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an MSD read buffer for signal generation enabling accurate quantification of inactive GLP-1 in mouse samples