Learn More
Meso Scale Discovery UPLEX MSE GLP1 inactive ASY
SDP
Catalog No. 5000082068
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for measuring GLP-1 (inactive) in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the Meso QuickPlex SQ 120 The assay detects 50-10 000 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium labels calibrators and controls enabling high-throughput screening and pharmacodynamic monitoring with low sample volume
Catalog No. 50-000-82068 Supplier Meso Scale Discovery Supplier No. K1525VK1
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for measuring GLP-1 (inactive) in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on the Meso QuickPlex SQ 120 The assay detects 50-10 000 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium labels calibrators and controls enabling high-throughput screening and pharmacodynamic monitoring with low sample volume