Learn More
Meso Scale Discovery UPLEX MSE GLP1 inactive ASYQP
SDP
Catalog No. 5000079383
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of inactive GLP-1 The assay has a lower limit of detection of 1 15 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours allowing high-throughput analysis This immunoassay provides a useful tool for studying GLP-1 biology and metabolism
Catalog No. 50-000-79383 Supplier Meso Scale Discovery Supplier No. K1525VK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of inactive GLP-1 The assay has a lower limit of detection of 1 15 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours allowing high-throughput analysis This immunoassay provides a useful tool for studying GLP-1 biology and metabolism