Learn More
Meso Scale Discovery UPLEX MSE GLP1 inactive ASYQP
SDP
Catalog No. 5000082069
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Inactive Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of inactive GLP-1 in mouse samples such as serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 3 5 hour turnaround time from sample to results The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-82069 Supplier Meso Scale Discovery Supplier No. K1525VK21
The Meso Scale Discovery U-PLEX Mouse GLP-1 Inactive Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of inactive GLP-1 in mouse samples such as serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay can be fully automated and has a rapid 3 5 hour turnaround time from sample to results The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance