Learn More
Meso Scale Discovery UPLEX MSE GLP1 inactive ASYQP
SDP
Catalog No. 5000082172
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The assay can detect inactive GLP-1 over a wide dynamic range of 78-20 000 pg/mL This product requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for analysis Overall the U-PLEX Mouse GLP-1 (Inactive) Assay provides a robust and efficient system for measuring inactive GLP-1 levels in mouse samples
Catalog No. 50-000-82172 Supplier Meso Scale Discovery Supplier No. K1525VK22
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of inactive GLP-1 in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The assay can detect inactive GLP-1 over a wide dynamic range of 78-20 000 pg/mL This product requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for analysis Overall the U-PLEX Mouse GLP-1 (Inactive) Assay provides a robust and efficient system for measuring inactive GLP-1 levels in mouse samples