Learn More
Meso Scale Discovery UPLEX MSE GLP1 TTL ASY
SDP
Catalog No. 5000079386
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with capture antibody to bind total GLP-1 Detection antibody conjugated to an electrochemiluminescent label (MSD SULFO-TAG) binds to the captured analyte Addition of read buffer induces electrochemiluminescence which is measured using the MSD instrument platform The kit includes a calibrator to generate a standard curve and has a detection range of 1 5-1000 pg/mL GLP-1 with an LLOQ of 1 5 pg/mL
Catalog No. 50-000-79386 Supplier Meso Scale Discovery Supplier No. K1525UK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with capture antibody to bind total GLP-1 Detection antibody conjugated to an electrochemiluminescent label (MSD SULFO-TAG) binds to the captured analyte Addition of read buffer induces electrochemiluminescence which is measured using the MSD instrument platform The kit includes a calibrator to generate a standard curve and has a detection range of 1 5-1000 pg/mL GLP-1 with an LLOQ of 1 5 pg/mL