Learn More
Meso Scale Discovery UPLEX MSE GLPactive ASY384
SDP
Catalog No. 5000081210
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid 3 5 hour single-step immunoassay with high sensitivity and wide dynamic range This assay kit includes a 384-well high bind plate detection antibody calibrator and assay diluent to enable quantification of active GLP-1 between 0 15-2500 pg/mL
Catalog No. 50-000-81210 Supplier Meso Scale Discovery Supplier No. K2526LK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in mouse serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid 3 5 hour single-step immunoassay with high sensitivity and wide dynamic range This assay kit includes a 384-well high bind plate detection antibody calibrator and assay diluent to enable quantification of active GLP-1 between 0 15-2500 pg/mL