Learn More
Meso Scale Discovery UPLEX MSE GLPactive ASY384well
SDP
Catalog No. 5000081274
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active GLP-1 in mouse samples such as serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results It includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L
Catalog No. 50-000-81274 Supplier Meso Scale Discovery Supplier No. K2526LK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Active) 384-Well Assay Kit is a multiplex immunoassay that measures active GLP-1 in mouse samples such as serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results It includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L