Learn More
Meso Scale Discovery UPLEX MSE GLPinactive ASY384
SDP
Catalog No. 5000079981
Encompass
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) 384-Well Assay Kit measures mouse GLP-1 (inactive) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array/multi-spot plate technology for sensitive detection The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and buffer for 384 tests Results are available in 3 5 hours using an MSD instrument for 384-well plates offering a sensitive and reproducible method to quantify mouse GLP-1 (inactive)
Catalog No. 50-000-79981 Supplier Meso Scale Discovery Supplier No. K2525VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GLP-1 (Inactive) 384-Well Assay Kit measures mouse GLP-1 (inactive) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array/multi-spot plate technology for sensitive detection The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and buffer for 384 tests Results are available in 3 5 hours using an MSD instrument for 384-well plates offering a sensitive and reproducible method to quantify mouse GLP-1 (inactive)