Learn More
Meso Scale Discovery UPLEX MSE Glucagon ASY
SDP
Catalog No. 5000081480
Encompass
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that enables simultaneous quantification of glucagon in mouse samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a mouse glucagon calibrator and a glucagon-specific antibody to detect and quantify glucagon levels between 0 08-500 pg/mL The assay requires a low sample volume of 50 L and provides high sensitivity wide dynamic range and minimal cross-reactivity The kit allows high-throughput analysis of glucagon in mouse serum plasma and cell culture supernatants
Catalog No. 50-000-81480 Supplier Meso Scale Discovery Supplier No. K1525YK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that enables simultaneous quantification of glucagon in mouse samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a mouse glucagon calibrator and a glucagon-specific antibody to detect and quantify glucagon levels between 0 08-500 pg/mL The assay requires a low sample volume of 50 L and provides high sensitivity wide dynamic range and minimal cross-reactivity The kit allows high-throughput analysis of glucagon in mouse serum plasma and cell culture supernatants