Learn More
Meso Scale Discovery UPLEX MSE Glucagon ASY384well
SDP
Catalog No. 5000079463
Encompass
The Meso Scale Discovery Mouse Glucagon U-PLEX Assay is a multiplex immunoassay that measures glucagon levels in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 The assay has a detection range of 1 53 - 10 000 pg/mL and the total assay time is approximately 3 5 hours This assay allows the simultaneous measurement of glucagon alongside other biomarkers
Catalog No. 50-000-79463 Supplier Meso Scale Discovery Supplier No. K2525YK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse Glucagon U-PLEX Assay is a multiplex immunoassay that measures glucagon levels in mouse biological samples such as serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 The assay has a detection range of 1 53 - 10 000 pg/mL and the total assay time is approximately 3 5 hours This assay allows the simultaneous measurement of glucagon alongside other biomarkers