Learn More
Meso Scale Discovery UPLEX MSE Glucagon ASY384well
SDP
Catalog No. 5000081270
Encompass
The Meso Scale Discovery Mouse Glucagon Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of glucagon in mouse samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay can detect glucagon concentrations as low as 0 3 pg/mL and has a standard curve range from 0 3-10 000 pg/mL The total assay time is 3 5 hours This assay kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-81270 Supplier Meso Scale Discovery Supplier No. K2525YK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse Glucagon Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of glucagon in mouse samples such as serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The assay can detect glucagon concentrations as low as 0 3 pg/mL and has a standard curve range from 0 3-10 000 pg/mL The total assay time is 3 5 hours This assay kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent