Learn More
Meso Scale Discovery UPLEX MSE Glucagon ASYQP
SDP
Catalog No. 5000082352
Encompass
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that allows simultaneous measurement of glucagon in mouse samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and diluents It can accurately quantify glucagon levels between 0 17-1500 pg/mL in mouse serum plasma and cell culture supernatants The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82352 Supplier Meso Scale Discovery Supplier No. K1525YK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Glucagon Assay Kit is a multiplex immunoassay that allows simultaneous measurement of glucagon in mouse samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and diluents It can accurately quantify glucagon levels between 0 17-1500 pg/mL in mouse serum plasma and cell culture supernatants The assay takes approximately 3 5 hours to complete