Learn More
Meso Scale Discovery UPLEX MSE GMCSF ASY384well
SDP
Catalog No. 5000081189
Encompass
The Meso Scale Discovery U-PLEX Mouse GM-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in mouse samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 1 3 pg/mL The assay takes approximately 3 5 hours to complete and includes plates detection antibodies calibrators and other necessary reagents
Catalog No. 50-000-81189 Supplier Meso Scale Discovery Supplier No. K252UMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse GM-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of GM-CSF in mouse samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 1 3 pg/mL The assay takes approximately 3 5 hours to complete and includes plates detection antibodies calibrators and other necessary reagents