Learn More
Meso Scale Discovery UPLEX MSE IFN ASY384well
SDP
Catalog No. 5000081283
Encompass
The Meso Scale Discovery Human IFN-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human interferon alpha (IFN-alpha) in various samples It uses MSD s SULFO-TAG labels and multi-array plate technology offering a detection limit of 0 04 pg/mL and a range up to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent It runs on MSD s SECTOR Imager for detecting and quantifying IFN-alpha levels
Catalog No. 50-000-81283 Supplier Meso Scale Discovery Supplier No. K252W1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human IFN-alpha Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for measuring human interferon alpha (IFN-alpha) in various samples It uses MSD s SULFO-TAG labels and multi-array plate technology offering a detection limit of 0 04 pg/mL and a range up to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent It runs on MSD s SECTOR Imager for detecting and quantifying IFN-alpha levels