Learn More
Meso Scale Discovery UPLEX MSE IFN ASYQP
SDP
Catalog No. 5000079053
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is a multiplex immunoassay for measuring mouse interferon alpha in various biological samples It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent It quantifies mouse IFN alpha levels accurately with high reproducibility and low cross-reactivity aiding immunology and inflammation research
Catalog No. 50-000-79053 Supplier Meso Scale Discovery Supplier No. K152W1K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IFN alpha Assay Kit is a multiplex immunoassay for measuring mouse interferon alpha in various biological samples It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and assay diluent It quantifies mouse IFN alpha levels accurately with high reproducibility and low cross-reactivity aiding immunology and inflammation research