Learn More
Meso Scale Discovery UPLEX MSE IFNB ASY384well
SDP
Catalog No. 5000081255
Encompass
The Meso Scale Discovery U-PLEX Mouse IFN beta 384-Well Assay Kit is designed to quantitatively measure mouse interferon beta in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IFN beta on the MESO QuickPlex SQ 120 instrument The kit includes a mouse IFN beta calibrator and 384-well plates pre-coated with capture antibody enabling high-throughput analysis of up to 80 samples
Catalog No. 50-000-81255 Supplier Meso Scale Discovery Supplier No. K252G0K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IFN beta 384-Well Assay Kit is designed to quantitatively measure mouse interferon beta in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IFN beta on the MESO QuickPlex SQ 120 instrument The kit includes a mouse IFN beta calibrator and 384-well plates pre-coated with capture antibody enabling high-throughput analysis of up to 80 samples