Learn More
Meso Scale Discovery UPLEX MSE IL10 ASY384well
SDP
Catalog No. 5000081208
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-10 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer It allows simultaneous measurement of IL-10 along with other biomarkers The assay takes 3 5 hours to complete
Catalog No. 50-000-81208 Supplier Meso Scale Discovery Supplier No. K252TZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-10 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 10 (IL-10) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and buffer It allows simultaneous measurement of IL-10 along with other biomarkers The assay takes 3 5 hours to complete