Learn More
Meso Scale Discovery UPLEX MSE IL12p70 ASY
SDP
Catalog No. 5000082154
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12p70 Assay Kit is designed for the quantitative measurement of mouse interleukin 12 (IL-12) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IL-12p70 The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse IL-12p70
Catalog No. 50-000-82154 Supplier Meso Scale Discovery Supplier No. K152UAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-12p70 Assay Kit is designed for the quantitative measurement of mouse interleukin 12 (IL-12) in serum plasma cell culture supernatants cell and tissue lysates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide sensitive and rapid detection of mouse IL-12p70 The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 2500 pg/mL for mouse IL-12p70