Learn More
Meso Scale Discovery UPLEX MSE IL12p70 ASY384well
SDP
Catalog No. 5000079978
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-12p70 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 12 (IL-12) in serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-12p70 quantitation with a detection limit of 1 3 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and optimized assay diluent The no-wash procedure enables rapid high-throughput cytokine profiling
Catalog No. 50-000-79978 Supplier Meso Scale Discovery Supplier No. K252UAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-12p70 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 12 (IL-12) in serum plasma and other samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-12p70 quantitation with a detection limit of 1 3 pg/mL The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and optimized assay diluent The no-wash procedure enables rapid high-throughput cytokine profiling