Learn More
Meso Scale Discovery UPLEX MSE IL15 ASY384well
SDP
Catalog No. 5000080183
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-15 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers It allows simultaneous measurement of IL-15 along with other biomarkers The assay takes approximately 3 hours to complete
Catalog No. 50-000-80183 Supplier Meso Scale Discovery Supplier No. K252URK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-15 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 15 (IL-15) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers It allows simultaneous measurement of IL-15 along with other biomarkers The assay takes approximately 3 hours to complete