Learn More
Meso Scale Discovery UPLEX MSE IL16 ASY384well
SDP
Catalog No. 5000080184
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-16 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a mouse IL-16 standard and a 384-well plate pre-coated with capture antibodies The assay has a detection range of 1 5-2500 pg/mL and exhibits minimal cross-reactivity Each kit provides sufficient reagents to perform 3 x 384-well plates when used as directed This product enables sensitive reproducible quantification of mouse IL-16
Catalog No. 50-000-80184 Supplier Meso Scale Discovery Supplier No. K252USK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-16 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 16 (IL-16) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a mouse IL-16 standard and a 384-well plate pre-coated with capture antibodies The assay has a detection range of 1 5-2500 pg/mL and exhibits minimal cross-reactivity Each kit provides sufficient reagents to perform 3 x 384-well plates when used as directed This product enables sensitive reproducible quantification of mouse IL-16