Learn More
Meso Scale Discovery UPLEX MSE IL17A ASY384well
SDP
Catalog No. 5000079321
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 17A (IL-17A) in various biological samples It uses MSD s electrochemiluminescent detection technology and is compatible with MSD s 384-well MULTI-ARRAY and SECTOR instruments The kit includes a mouse IL-17A calibrator and a pre-coated plate with capture antibodies It has a detection range of 0 05-1000 pg/mL and minimal cross-reactivity with other cytokines enabling sensitive and accurate quantification of mouse IL-17A for research
Catalog No. 50-000-79321 Supplier Meso Scale Discovery Supplier No. K252UTK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17A 384-Well Assay Kit is a multiplex immunoassay for measuring mouse interleukin 17A (IL-17A) in various biological samples It uses MSD s electrochemiluminescent detection technology and is compatible with MSD s 384-well MULTI-ARRAY and SECTOR instruments The kit includes a mouse IL-17A calibrator and a pre-coated plate with capture antibodies It has a detection range of 0 05-1000 pg/mL and minimal cross-reactivity with other cytokines enabling sensitive and accurate quantification of mouse IL-17A for research