Learn More
Meso Scale Discovery UPLEX MSE IL17A ASYQP
SDP
Catalog No. 5000078475
Encompass
The U-PLEX Mouse IL-17A Assay (K152UTK) is a highly sensitive electrochemiluminescence immunoassay designed to quantify interleukin-17A levels in mouse biological samples This multiplex-ready assay offers superior sensitivity compared to traditional ELISA methods enabling detection of low-abundance cytokines in small sample volumes The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammatory responses and immune system function
Catalog No. 50-000-78475 Supplier Meso Scale Discovery Supplier No. K152UTK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse IL-17A Assay (K152UTK) is a highly sensitive electrochemiluminescence immunoassay designed to quantify interleukin-17A levels in mouse biological samples This multiplex-ready assay offers superior sensitivity compared to traditional ELISA methods enabling detection of low-abundance cytokines in small sample volumes The kit includes pre-coated plates detection antibodies and calibration standards for reliable reproducible results in research applications studying inflammatory responses and immune system function