Learn More
Meso Scale Discovery UPLEX MSE IL17A/F ASY
SDP
Catalog No. 5000080451
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A and 17F in various samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range The kit includes a 96-well plate with necessary antibodies calibrators and controls It processes up to 80 samples in 3 5 hours enabling efficient analysis of IL-17A and IL-17F crucial in autoimmunity and inflammation
Catalog No. 50-000-80451 Supplier Meso Scale Discovery Supplier No. K152VYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A and 17F in various samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range The kit includes a 96-well plate with necessary antibodies calibrators and controls It processes up to 80 samples in 3 5 hours enabling efficient analysis of IL-17A and IL-17F crucial in autoimmunity and inflammation