Learn More
Meso Scale Discovery UPLEX MSE IL17A/F ASY384well
SDP
Catalog No. 5000081193
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It processes up to 80 samples in 3 5 hours ensuring efficient sensitive and reproducible IL-17A and IL-17F measurement
Catalog No. 50-000-81193 Supplier Meso Scale Discovery Supplier No. K252VYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17A/F Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in various biological matrices It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It processes up to 80 samples in 3 5 hours ensuring efficient sensitive and reproducible IL-17A and IL-17F measurement