Learn More
Meso Scale Discovery UPLEX MSE IL17C ASY384well
SDP
Catalog No. 5000079323
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s SULFO-TAG labels and multi-array/multi-spot plate technology for sensitive reproducible results from small samples It detects 0 05-1000 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium buffers blocker solutions calibrator and controls enabling high throughput quantitative IL-17C analysis with minimal sample use
Catalog No. 50-000-79323 Supplier Meso Scale Discovery Supplier No. K252WJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17C Assay is a multiplex immunoassay kit for quantifying mouse interleukin 17C (IL-17C) in serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s SULFO-TAG labels and multi-array/multi-spot plate technology for sensitive reproducible results from small samples It detects 0 05-1000 pg/mL and includes a plate with capture antibodies detection antibodies with ruthenium buffers blocker solutions calibrator and controls enabling high throughput quantitative IL-17C analysis with minimal sample use