Learn More
Meso Scale Discovery UPLEX MSE IL17C ASYQP
SDP
Catalog No. 5000080366
Encompass
The U-PLEX Mouse IL-17C Assay (K152WJK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of interleukin-17C protein levels in mouse biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements The kit provides researchers with validated reagents and protocols for cytokine analysis in mouse models supporting inflammation and immunology research applications
Catalog No. 50-000-80366 Supplier Meso Scale Discovery Supplier No. K152WJK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse IL-17C Assay (K152WJK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of interleukin-17C protein levels in mouse biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements The kit provides researchers with validated reagents and protocols for cytokine analysis in mouse models supporting inflammation and immunology research applications